Specifications
- Antigen name:PKC theta
- Antigen symbol:PRKCQ
- Clonality:Monoclonal
- Clone:HL1157
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rhesus monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Format:Azide and BSA free
- Antigen synonyms:PRKCT|nPKC-theta|MGC141919|MGC126514|PKC theta|protein kinase C theta type|EC 2.7.11|EC 2.7.11.13|protein kinase C, theta|p Thr219
- Modification:p Thr219
- Purity:Protein A purified.
- Storage buffer:PBS
- Molecular weight:82 kDa
- Storage temperature:Store at 4 ℃ short term. Aliquot and store at –20 ℃ long term. Avoid freeze-thaw cycles.
- Immunogen:Carrier-protein conjugated synthetic peptide surrounding phospho Thr219 of human PKC theta. The exact sequence is proprietary.
- Pk:100 µl
Specifications
About this item
Members of the protein kinase C (PKC) family play a key regulatory role in variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC.
PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms (alpha, beta I, beta II and gamma) and novel (n) PKC isoforms (delta, episilon, zeta, eta and theta). Patterns of expression for each PKC isoform differs among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and episilon are independent of Ca⁺⁺. On the other hand, nPKC and episilon, as well as all of the cPKC members, possess phorbol ester-binding activities and kinase activities.
This product is for research use only and is not approved for use in humans or in clinical diagnosis. Primary Antibodies are guaranteed for one year from date of receipt.
Type:
Antigen: PRKCQ
Clonality: Monoclonal
Clone: HL1157
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human