Pig Pepsin (from Gastric mucosa)
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:9001-75-6
- Source:Gastric mucosa
- Species:Pig
- Storage conditions:Refrigerator
- Enzyme name:Pepsin
Specifications
Frequently Bought Together