Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-bromopyrimidine
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine 97%
Irritant
2-Amino-5-bromopyrimidine 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Amino-5-bromopyrimidine 97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
7752-82-1
:
Formula:
C₄H₄BrN₃
MDL Number:
MFCD00012341
:
174 g/mol
Melting Pt:
232…234 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAH27291
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation