Order Entry
Norway
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol ≥99%

3-Fluoropyrocatechol ≥99%

Corrosive
Irritant
3-Fluoropyrocatechol ≥99%
Certificates
3-Fluoropyrocatechol ≥99%
3-Fluoropyrocatechol ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 363-52-0 Molecular Formula: C6H5FO2 Formula Weight: 128.10 Melting Point: 71-73° Beilstein Reference: 2436074 UN No.: 1759 MDL No.: MFCD00042582

Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:71…73 °C
  • Density:1.514 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - kr 710,00
    kr 710,00
    1 * 1 g
       kr 710,00

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - kr 2 075,00
    kr 2 075,00
    1 * 5 g
       kr 2 075,00