1,8-Naphthalimide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:81-83-4
:
Formula:C₁₂H₇NO₂
MDL Number:MFCD00006920
:197.19 g/mol
Boiling Pt:475 °C (1013 hPa)
Melting Pt:299…303 °C
Density:1.482 g/cm³ (19.85 °C)
Storage Temperature:Ambient
EINECS:201-379-7