

4-Formylbenzoic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:619-66-9
:
Formula:C₈H₆O₃
MDL Number:MFCD00006951
:150.13 g/mol
Boiling Pt:306 °C (1013 hPa)
Melting Pt:247 °C
EINECS:210-607-4
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥95 %
- NMR (1H-NMR):Consistent with structure