Order Entry
Norway
ContactUsLinkComponent
 
 
2-Formylbenzoic acid 99%
 
undefined
 
2-Formylbenzoic acid 99%

 

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    96 to 99 °C
  • HPLC:
    ≥98.5 %