9-Hydroxyfluorene 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
9-Hydroxyfluorene 97%
Catalog # ACRO121230250
Supplier: Thermo Fisher Scientific
CAS Number: 1689-64-1
Jump to:
Product Details & Documents
Synonyms: 9-Fluorenol, Fluoren-9-ol
- CAS Number:1689-64-1
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00001135
- Molecular Weight:182.22 g/mol
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:152…158 °C
- Density:1,151 g/cm³ (20 °C)
- EINECS:216-879-0
Documents
ACRO12123
SDS
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:152 to 158 °C
- GC:≥96.0 %
- Titration with HClO4:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



