Order Entry
Norway
Orders LinkContactUsLinkComponent
Demonstration instruments for measuring inserts
Marketsource
Supplier:   PHYWE
Demonstration instruments for measuring inserts
Supplier:  PHYWE
Marketsource
Certificates
Demonstration instruments for measuring inserts
Demonstration instruments for measuring inserts
42 Options Available Below
Product Details & Documents
With a special addition, the analogue instrument can be turned into a digital instrument with simultaneous analogue display. In order to extend the measurement possibilities, there are also measurements for power and voltage amplification. The replaceable measuring areas are slid into a slot on the top of the instrument ('toaster principle').
The operation can be performed by hand. Special measurements eg for Resistance and Lux is inserted horizontally from the back. All switches are done automatically via a 16-pin connection list. Most measuring ranges are passive and are therefore independent of batteries or network connection.
The scales are large and clear with fingering and completely linear dividing even for alternating current. Reading can also be done from the back because the scales are double-sided and have a gap. Zero adjustment of the pointer is performed mechanically with thumbwheel on the underside of the instrument. The instrument is equipped with extensive overload protection through diodes, cold conductors and fuses. In addition, the active measuring areas are electronically protected.
Product Family Options
Current measurement, DC
Product Information
  • Range
    Availability
    Price
  • 1mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 1 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 1-0-1 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 µA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 µA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 30 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
Voltage measurement, DC
Product Information
  • Range
    Availability
    Price
  • 30 mV
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 mV
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 mV
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    Marketsource
    Marketsource
  • 1 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 30 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 kV
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 1 kV
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
Current measurement, AC
Product Information
  • Range
    Availability
    Price
  • 1 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 1 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 A
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 µA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 µA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 30 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 mA
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
Voltage measurement, AC
Product Information
  • Range
    Availability
    Price
  • 1 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 3 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 10 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 30 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 100 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
  • 300 V
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
Storage rack
Product Information
  • Range
    Availability
    Price
  • 765-1248
    Storage rack for measuring areas
    Storage rack for measuring areas
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource
Product Information
  • Range
    Availability
    Price
  • 765-1247
    Demonstration instruments for measuring inserts
    Demonstration instruments for measuring inserts
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    Marketsource
    Marketsource