Order Entry
Norway
ContactUsLinkComponent
Panel Mechanics 2, completion kit
Panel Mechanics 2, completion kit
Catalog # 765-1150
Supplier:  PHYWE
undefined
Panel Mechanics 2, completion kit
Catalog # 765-1150
Supplier:  PHYWE
Specifications
  • Description:
    Panel Mechanics 2, completion kit
  • Pk:
    1
Specifications
About this item
Supplementary kit for Painting Mechanics 1. Contains 11 components with which a further 18 experiments can be performed in the areas of motion, energy forms and the mechanics of liquids and gases. The experiments assume that components are used from the Tavelmekanik 1 kit. Examples of experiments:
  • Uniform rectilinear motion.
  • Horizontal and angled throw.
  • Newton's foundations.
  • Energy conversion when traveling on a roller coaster.
  • Communicating vessels.
  • Hydraulic pressure.
  • Archimedes' principle.
  • Pressure in flowing liquid.
  • Pressure in gases.
  • Boyle-Mariotte's team