Order Entry
Norway
ContactUsLinkComponent
Whiteboard mechanics 1, basic set with whiteboard
Whiteboard mechanics 1, basic set with whiteboard
Catalog # 765-1149
Supplier:  PHYWE
undefined
Whiteboard mechanics 1, basic set with whiteboard
Catalog # 765-1149
Supplier:  PHYWE
Specifications
  • Description:
    Whiteboard mechanics 1, basic set with whiteboard
  • Pk:
    1
Specifications
About this item
Contains 50 different components including detailed instructions. Delivered in a plywood box, 31 different attempts are described in the manual, e.g. Examples of experiments:
  • Power parallelogram
  • Hooks laugh
  • Leaning planet
  • Composition of forces
  • Lever
  • Trundle
  • Lifting block
  • Pendulum
  • Torque
  • Friction