Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(3-Bromo-5-chlorophenyl)acetic acid
2-(3-Bromo-5-chlorophenyl)acetic acid 95%
2-(3-Bromo-5-chlorophenyl)acetic acid 95%
Irritant
2-(3-Bromo-5-chlorophenyl)acetic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(3-Bromo-5-chlorophenyl)acetic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
202001-01-2
:
Formula:
C₈H₆BrClO₂
MDL Number:
MFCD18373986
:
249.49 g/mol
Melting Pt:
120…122 °C
:
GHS07