Order Entry
Norway
ContactUsLinkComponent
 
Organic reference standard, Ethyl acetate 2,000 µg/ml in methanol, Supelco®
  47947.
 :  Merck
 :  
Organic reference standard, Ethyl acetate 2,000 µg/ml in methanol, Supelco®
  47947.
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    141-78-6
  • Description:
    Ethyl acetate 2,000 µg/ml in methanol
  • Formula:
    C₄H₈O₂
  • Description:
    Ethyl acetate