Saponin
Catalog # ROTH6857.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
Density:1.015…1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-462-6
CAS Number:8047-15-2
MDL Number:MFCD00081981