Order Entry
Norway
ContactUsLinkComponent
Saponin
Saponin
Catalog # ROTH4185.1
Supplier:  Roth Carl
CAS Number:  
Saponin
Catalog # ROTH4185.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
Density:1.015…1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-462-6
CAS Number:8047-15-2
MDL Number:MFCD00081981