Order Entry
Norway
ContactUsLinkComponent
 
Saponin (from Quillaja Saponaria Molina) pract.
 
undefined
Saponin (from Quillaja Saponaria Molina) pract.
 

 

  • Appearance:
    Dark red to tan Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (20 % in water) Clear to hazy foaming solution
  • Moisture:
    ≤10 %
  • Ash:
    ≤10 %
  • pH:
    3.8 to 4.6 (20 % solution)