Order Entry
Norway
ContactUsLinkComponent
5(6)-ROX N-succinimidyl ester mixture of isomers ≥90%
5(6)-ROX N-succinimidyl ester mixture of isomers ≥90%
Catalog # AATB390
Supplier:  AAT Bioquest
CAS Number:  
undefined
5(6)-ROX N-succinimidyl ester mixture of isomers ≥90%
Catalog # AATB390
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :631.67 g/mol
CAS Number:114616-32-9
Formula:C₃₇H₃₃N₃O₇