Order Entry
Norway
ContactUsLinkComponent
Biotin hydrazide, ReadiView™ biodetection
Irritant
Biotin hydrazide, ReadiView™ biodetection
Catalog # AATB3055
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Biotin hydrazide, ReadiView™ biodetection
Catalog # AATB3055
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :258.34 g/mol
ADR:5,II
 :GHS07
CAS Number:66640-86-6
Formula:C₁₀H₁₈N₄O₂S
MDL Number:MFCD00078532