Order Entry
Norway
ContactUsLinkComponent
Acridine Orange 10-nonyl bromide (NAO) ≥95%
Irritant
Acridine Orange 10-nonyl bromide (NAO) ≥95%
Catalog # AATB22205
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Acridine Orange 10-nonyl bromide (NAO) ≥95%
Catalog # AATB22205
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :472.50 g/mol
ADR:5,II
 :GHS07
CAS Number:75168-11-5
Formula:C₂₆H₃₈BrN₃
MDL Number:MFCD00467605