5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥90%, ReadiUse™
Catalog # AATB22028
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2.500 µG
- Seq:2175
Specifications
About this item
:
:557.467 g/mol
ADR:5,II
:GHS07
CAS Number:150347-59-4
:MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466