Order Entry
Norway
ContactUsLinkComponent
OG488-Tris NTA-Ni complex, HIS Lite™
Irritant
OG488-Tris NTA-Ni complex, HIS Lite™
Catalog # AATB12615
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
OG488-Tris NTA-Ni complex, HIS Lite™
Catalog # AATB12615
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
ADR:5,II
 :GHS07