Generator with hand loom
Generator with hand loom
765-0210
Marketsource
Bicycle dynamo with transparent housing. Consists of magnetic anchor and coil mounted on base with belt drive. Equipped with connections for 4 mm banana plug and light bulb 3.5 V/0.2 A.
- Generator: 6 V / 3 W
- Base dimensions: 120 × 90 × 23 mm
- Description:Generator with hand loom