Order Entry
Norway
ContactUsLinkComponent
Heat radiation
Heat radiation
Catalog # 765-0177
undefined
Heat radiation
Catalog # 765-0177
Specifications
  • Description:
    Heat radiation
  • Pk:
    1
Specifications
About this item
For experiments with absorption of heat radiation. Consists of a black and a white E-flask and two thermometers (0 to 50 °C).
  • Comes with insulating cork plate