Order Entry
Norway
ContactUsLinkComponent
Fluorescein diacetate ≥95%
Fluorescein diacetate ≥95%
Catalog # AATB22020
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Fluorescein diacetate ≥95%
Catalog # AATB22020
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :416.38 g/mol
ADR:5,II
 :GHS07
CAS Number:596-09-8
Formula:C₂₄H₁₆O₇
MDL Number:MFCD00005062