Table optics, TESS OE3
Catalog # 765-1319
Supplier: PHYWE
Specifications
- Description:TESS Optics OE3
- Pk:1
Specifications
About this item
TESS OE3 contains 19 components. With TESS OE3 as a complement to TESS OE1 and OE2, another 30 experiments can be performed in the field of wave optics. Delivered in a plywood box.