Order Entry
Norway
ContactUsLinkComponent
Laser with magnetic mount
Laser with magnetic mount
Catalog # 765-1003
Supplier:  KVANT LIFE SCIENCE
undefined
Laser with magnetic mount
Catalog # 765-1003
Supplier:  KVANT LIFE SCIENCE
Specifications
  • Description:
    Laser with magnetic mount, red
  • Pk:
    1
Specifications
About this item
Laser light source for mounting on magnetic board (white board or similar). Can be used to advantage in combination with picture optics (765-1009).
  • Several lasers can be mounted in parallel if a beam is desired