Order Entry
Norway
Orders LinkContactUsLinkComponent
UV screen
UV screen
UV screen
Catalog # 765-0161
Marketsource
UV screen
Catalog # 765-0161
Marketsource
UV screen
UV screen
UV screen
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
To display ultraviolet radiation. Half of the plate is coated with a fluorescent substance that emits light during UV radiation.
Dimensions: 150 × 100 mm
Specifications
  • Description:
    UV screen