Fluorescein disodium salt 1% in aqueous solution
Catalog # 765-0930
Supplier: GR NNA PLASTEMBALLAGE AB
CAS Number:
Specifications
- Pack type:Glass dropper bottle
- Pk:30 mL
- Seq:2345
Specifications
About this item
:
:376.28 g/mol
CAS Number:518-47-8
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039