Goat Anti-Rat IgG2a Secondary Antibody
Katalog # NOVUNBP2-68486
Supplier: Novus Biologicals
Specifications
- Antibody type:Secondary
- Antigen name:IgG2a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:Yes
- Purity:Affinity purified
- Storage buffer:PBS
- Storage temperature:Store at 4 °C
- Concentration:1 mg/ml
- Size:0,5 mg
- Pk:0,5 mg
Specifications
About this item
Goat anti Rat IgG2a antibody recognises rat IgG2a as assessed by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG2a from other species.
Recommended Dilutions: ELISA: 1 to 10 µg/ml, Flow Cytometry, Western Blot
Preservative: 0.1% Sodium Azide
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat