2,4,6-Tribromoanisole
Catalog # CMPLSB19790.100MG
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- XLogP3:4.1
- Hydrogen bond donor count:0
- Rotatable bond count:0
- Exact mass:1
- Monoisotopic mass:343.78700 g/mol
- Topological polar surface area:341.78905 g/mol
- Heavy atom count:9.2 Ų
- Formal charge:11
- Complexity:0
- Isotope atom count:119
- Defined atom stereocenter count:0
- Undefined atom stereocenter count:0
- Defined bond stereocenter count:0
- Undefined bond stereocenter count:0
- Covalently-bonded unit count:1
- Compound is canonicalized:Yes
- Hydrogen bond acceptor count:1
Specification Test Results
About this item
:
:344.83 g/mol
Boiling Pt:298 °C
Melting Pt:88 °C
Density:25 °C
:GHS09
CAS Number:607-99-8
Formula:C₇H₅Br₃O
MDL Number:MFCD00192510