Anti-JAK3 Rabbit Recombinant Antibody [clone: JAK3Y980981-E10] (PE)
Catalog # ABNORAB02867.100
Supplier: Abnova
Specifications
- Antigen symbol:JAK3
- Clonality:Recombinant
- Clone:JAK3Y980981-E10
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human JAK3.
- Pk:100 AU