Order Entry
Norway
ContactUsLinkComponent
 
Anti-UBE3A Rabbit Recombinant Antibody [clone: R02-1A0]
  ABNORAB01336.100UL
 :  Abnova
Anti-UBE3A Rabbit Recombinant Antibody [clone: R02-1A0]
  ABNORAB01336.100UL
 :  Abnova

 

  • Antigen symbol:
    UBE3A
  • Clonality:
    Recombinant
  • Clone:
    R02-1A0
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human UBE3A.
  • Pk:
    100 µl