Order Entry
Norway
ContactUsLinkComponent
Anti-IHH Rabbit Recombinant Antibody [clone: R03-3F2]
Anti-IHH Rabbit Recombinant Antibody [clone: R03-3F2]
Catalog # ABNORAB01426.100UL
Supplier:  Abnova
Anti-IHH Rabbit Recombinant Antibody [clone: R03-3F2]
Catalog # ABNORAB01426.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    IHH
  • Clonality:
    Recombinant
  • Clone:
    R03-3F2
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human IHH.
  • Pk:
    100 µl

Specifications

About this item