Order Entry
Norway
ContactUsLinkComponent
Accessories for Immersion thermostats, KISS E
Accessories for Immersion thermostats, KISS E
Catalog # HUBE10688
Supplier:  HUBER
Accessories for Immersion thermostats, KISS E
Catalog # HUBE10688
Supplier:  HUBER
Specifications
  • Description:
    Connection socket (Lemosa) for connecting a Pt100 measuring sensor
  • Pk:
    1
  • For:
    For Huber units with KISS controller