Electrophoresis power supplies, EVT 300
Catalog # 732-1424
Supplier: EDVOTEK
Specifications
- Description:EVT 300 power supply, 75 V / 150 V for 1 or 2 units
- Pk:1
Specifications
About this item
EVT 300 power supplies can operate M6Plus units, two M12 units or two HexaGels (at 75 V). The EVT 300 runs gels as fast as 20 to30 minutes (at 150 V).