About this item
Hei-PLATE Mix 'n' Heat Ultimate is equipped with numerous interfaces for automated laboratory processes. The RS232 and USB B ports allow a control via our integrated Hei-PROCESS solution.
The USB A interface at the front allows data logging via a USB stick. For more comfort and safety, you can connect the magnetic stirrer with the electric Heidolph lab jack via the USB C interface. There are three heating modes for temperature control: FAST for a quick heating, as well as PRECISE and PRECISE+ for the exact control of your target temperature without overshoot - this is how sensitive samples are optimally protected.
Therefore, the device is compatible with the temperature sensor Pt1000 and the double sensor Pt1000 that allows the simultaneous measurement in your sample and in the bath liquid. The possibility to program ramps and to create individual profiles, increases the precision and reproducibility of your experiments. For the stirring speed, there is a three-stage setting (start-up behavior). For mixing your sample optimally, you can set the rotational direction of the magnetic stirring bar and benefit from the automatic stir bar detector.
This magnetic stirrer offers all relevant safety features. The large safety distance between the heating plate and the control panel as well as the visual residual heat indicator at temperatures >50 °C protect against burns.
Specifications
- Catalog No:
- HEID506-41158-00
- HEID506-42100-00
- HEID506-41151-00
- HEID506-41150-00
- HEID506-41100-00
- Max. stirring capacity H₂O:
- 20 L
- 20 L
- Electrical:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Speed display:
- 2×TFT display
- 2×TFT display
- Plate material:
- Kera-Disk®, ceramic coated Silumin (aluminium alloy)
- Kera-Disk®, ceramic coated Silumin (aluminium alloy)
- Plate dimensions:
- 135 mm
- 145 mm
- Weight:
- 3 kg
- 3 kg
- Overall dimensions:
- 168×299×106 mm
- 168×299×106 mm