Order Entry
Norway
ContactUsLinkComponent
Hygen™ microfibre dust mop
Hygen™ microfibre dust mop
Catalog # 141-0117
undefined
Hygen™ microfibre dust mop
Catalog # 141-0117

Specifications

  • Colour:
    Green
  • Description:
    HYGEN™ microfibre dust pad for dust, dirt, and debris
  • Dimensions:
    444×142×15 mm
  • Seq:
    04
  • Pk:
    1

Specifications

About this item

The Rubbermaid Commercial HYGEN™ Microfibre Dust Pads trap and lock away dust, dirt, and debris mechanically and electrostatically with split-blended fibres.

  • Dense 3 - 5 micrometer quality Fibre removes more dust, dirt and microbes than conventional cleaning products
  • Advanced hook-and-loop backing holds-up under harsh laundering conditions and attaches pads more securely
  • Double finished edges retain mop shape with use over time
  • Durable for up to 500 launderings (up to 200 with chlorine)
  • Chlorine safe in hot and cold water

For use with Hygen™ PULSE™ mops.