About this item
Measure pH or ORP simultaneously with mV and temperature using the Orion Lab Star PH111 pH/mV meter. Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is ideal for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. View active pH, ORP and temperature calibration logs with date and time stamp, record key measurements using the 500-point data log with date and time stamp, and output all logs to a printer or computer.
- Budget-friendly pH meter for simplified pH and ORP testing
- Display shows pH, mV and temperature simultaneously for accurate data recording
- Large 5 inch backlit display makes viewing and recording measurements easy
- Small meter footprint saves valuable bench space in your lab
- Onscreen stabilising and ready indicators help identify when readings are complete
- pH electrode condition icon helps ensure reliable electrode performance
- Date and time data stamped data and calibration logs support GLP compliance
- Use 500-point data log with date and time stamp to record key measurements
- Output data and calibration logs to a printer or computer for long-term data storage
- Three year meter warranty
- Backlight option is available
Measure −2 to +18 pH, −2000 to +2000 mV and −5 to +105 °C or 23 to 221 °F to ensure comprehensive measurement capabilities. Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance. View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements.
It is the pH meter and stand option, which allows you to purchase your pH electrode separately. The standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use.
It is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore. The low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate. It is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges.
AC Adapter: Universal 100 to 240 V 50/60 Hz power supply with twist-and-lock meter connection and interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Data Management Details: Transfer single or all data points to printer or computer
Data Points: 500 with Date and Time Stamp
Measurement Modes: pH with mV and temperature or ORP with mV and temperature
Log Function Type: Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Range (ORP): ±2000 mV with ±250 mV offset
Temperature Selection: Manual or automatic with ATC temperature probe
Specifications
- Catalog No:
- 662-2471
- 662-2470
- 662-2474
- 662-2472
- 662-2473
- pH range:
- –2,000...18,000 pH
- –2,000...18,000 pH
- –2,000...18,000 pH
- –2,000...18,000 pH
- –2,000...18,000 pH
- pH resolution:
- 0,1, 0,01, 0,001 pH
- 0,1, 0,01, 0,001 pH
- 0,1, 0,01, 0,001 pH
- 0,1, 0,01, 0,001 pH
- 0,1, 0,01, 0,001 pH
- pH accuracy:
- ±0,005 pH ±1 LSD (relative)
- ±0,005 pH ±1 LSD (relative)
- ±0,005 pH ±1 LSD (relative)
- ±0,005 pH ±1 LSD (relative)
- ±0,005 pH ±1 LSD (relative)
- pH calibration:
- 1 - 5 points
- 1 - 5 points
- 1 - 5 points
- 1 - 5 points
- 1 - 5 points
- mV range:
- ±2000 mV (mV/ORP)
- ±2000 mV (mV/ORP)
- ±2000 mV (mV/ORP)
- ±2000 mV (mV/ORP)
- ±2000 mV (mV/ORP)
- mV resolution:
- 0,1 mV (mV/ORP)
- 0,1 mV (mV/ORP)
- 0,1 mV (mV/ORP)
- 0,1 mV (mV/ORP)
- 0,1 mV (mV/ORP)
- mV accuracy:
- ±0,2 mV or ±0,05 % of the reading, whichever is greater (mV/ORP, relative)
- ±0,2 mV or ±0,05 % of the reading, whichever is greater (mV/ORP, relative)
- ±0,2 mV or ±0,05 % of the reading, whichever is greater (mV/ORP, relative)
- ±0,2 mV or ±0,05 % of the reading, whichever is greater (mV/ORP, relative)
- ±0,2 mV or ±0,05 % of the reading, whichever is greater (mV/ORP, relative)
- mV calibration:
- 1 point (mV/ORP)
- 1 point (mV/ORP)
- 1 point (mV/ORP)
- 1 point (mV/ORP)
- 1 point (mV/ORP)
- Temperature range:
- −5,0...105,0 °C (23,0...221,0 °F)
- −5,0...105,0 °C (23,0...221,0 °F)
- −5,0...105,0 °C (23,0...221,0 °F)
- −5,0...105,0 °C (23,0...221,0 °F)
- −5,0...105,0 °C (23,0...221,0 °F)
- Temperature resolution:
- 0,1 °C, 0,1 °F
- 0,1 °C, 0,1 °F
- 0,1 °C, 0,1 °F
- 0,1 °C, 0,1 °F
- 0,1 °C, 0,1 °F
- Temperature accuracy:
- ±0,3 °C, ±0,5 °F (relative)
- ±0,3 °C, ±0,5 °F (relative)
- ±0,3 °C, ±0,5 °F (relative)
- ±0,3 °C, ±0,5 °F (relative)
- ±0,3 °C, ±0,5 °F (relative)
- Temperature calibration:
- 1 point
- 1 point
- 1 point
- 1 point
- 1 point
- Display:
- 5" backlit LCD
- 5" backlit LCD
- 5" backlit LCD
- 5" backlit LCD
- 5" backlit LCD
- Power supply:
- 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1,3 A
- 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1,3 A
- 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1,3 A
- 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1,3 A
- 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1,3 A
- Data logging - log function:
- 500 sets with time and date
- 500 sets with time and date
- 500 sets with time and date
- 500 sets with time and date
- 500 sets with time and date
- Memory:
- Non-volatile
- Non-volatile
- Non-volatile
- Non-volatile
- Non-volatile
- IP rating:
- IP 54
- IP-54
- IP-54
- IP-54
- IP-54
- Outputs:
- Computer or printer (STARA-106)
- Computer or printer
- Computer or printer
- Computer or printer
- Computer or printer
- Input:
- BNC for pH or ORP, 8-pin MiniDIN for ATC
- BNC, ATC
- BNC, ATC
- BNC, ATC
- BNC, ATC
- Alarm:
- Yes, 1 - 168 hours or off (calibration alarm)
- Yes, 1 - 168 hours or off (calibration alarm)
- Yes, 1 - 168 hours or off (calibration alarm)
- Yes, 1 - 168 hours or off (calibration alarm)
- Yes, 1 - 168 hours or off (calibration alarm)
- Calibration:
- Automatic buffer recognition with manual buffer entry option
- Automatic buffer recognition with manual buffer entry option
- Automatic buffer recognition with manual buffer entry option
- Automatic buffer recognition with manual buffer entry option
- Automatic buffer recognition with manual buffer entry option
- W×D×H:
- 155,2×198,3×61,7 mm
- 155,2×198,3×61,7 mm
- 155,2×198,3×61,7 mm
- 155,2×198,3×61,7 mm
- 155,2×198,3×61,7 mm
- Weight:
- 700 g (1,54 lb.)
- 700 g (1,54 lbs.)
- 700 g (1,54 lbs.)
- 700 g (1,54 lbs.)
- 700 g (1,54 lbs.)