Replacement Air Filter for ULT Freezers
Catalog # HERA332297H02
Supplier: Thermo Scientific
Specifications
- Description:Replacement air filter chest freezers (quantity of 5)
- Pk:1
- For:TDEC39686FV
Specifications
About this item
Replacement air filter for ultra-low temperature chest freezers.
Routine maintenance is critical for proper ultra-low temperature (ULT) freezer operation. Replacing your ULT's old air filter enables the necessary airflow to the unit's refrigeration system.