About this item
The CYM17 Memosens-analog converter enables easy use of digital Memosens sensors in your fermentation applications in the laboratory without changing your SOP.
- Easy exchange of your existing analog sensors. Just install the Memosens sensors in your fermenter and connect them with the converter
- Thanks to Memosens technology, the cable connection is insensitive to moisture, corrosion or dirt: you can count on the sensor's highest availability and your test is safe
- Memosens sensors have an excellent performance in process applications as well as in benchtop fermenters
- Providing you with 100% measuring consistency from the first lab tests to the final scaled-up process
- The converter continuously checks the Memosens sensors for highest measuring reliability and sets off an alarm if any error occurs
The converter set for lab fermenters comprises a CYM17 converter, a pH sensor (CPS61E), an optical DO sensor (COS81E) and the appropriate CYK20 cables and the KSG8 standard adapter.
The CYM17 Memosens-analog converter enables easy use of digital Memosens sensors in your fermentation applications in the laboratory without changing your SOP. Memosens digital technology combines maximum process and data integrity with simple operation. It eliminates any moisture issues and guarantees stable signals after autoclavation.
The CYM17 enables the use of digital Memosens sensors in laboratories of biotechnology industries with your existing fermenters.
Converter set 2 for lab fermenters with 225 mm Memosens 2.0 sensors: Digital/analog converter for connection of Memosens pH and DO sensors to analog inputs; Memosens pH sensor; Memosens optical DO sensor; cables and adapter; CYM17-AA+SBUB (incl. 2×CYK20 plus KSG8 standard adapter); CPS61E-AA6NTP4JG; and COS81E-AA24ACLF1JG.