VWR®, Septa ND12
Katalog # 548-0185A
Supplier: VWR Collection
Specifications
- Septum:Universal HT injection port, transparent
- Thickness:3,0 mm
- Pk:25
Specifications
About this item
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
- Fits for a broad range of caps
- Easy exchange
- Different hardness and septa thickness available
Septa choice depends on the application.