Anti-GDPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9206R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GDPD3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:101-200/318
- Form:Liquid
- Gene ID:79153
- Antigen synonyms:GDPD3|GDE7|Glycerophosphodiester phosphodiesterase domain-containing protein 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GDPD3
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Hydrolyzes lysoglycerophospholipids to produce lysophosphatidic acid (LPA) and the corresponding amines. Shows a preference for 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF), lysophosphatidylcholine (lyso-PC) and N-acylethanolamine lysophospholipids. Does not display glycerophosphodiester phosphodiesterase activity, since it cannot hydrolyze either glycerophosphoinositol or glycerophosphocholine.gene mapping to human chromosome Xq13.1.
Type: Primary
Antigen: GDPD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/318
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat