BACH2 Overexpression Lysate (Adult Normal)
Catalog # NOVUNBL1-07899
Supplier: Novus Biologicals
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:100 µG
- Tested applications:WB
Specifications
About this item
BACH2 Overexpression Lysate (Adult Normal)
Tested Applications: Western Blot.