- Antibody type:Primary
- Antigen name:LPCAT2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:131-230/544
- Form:Liquid
- Gene ID:54947
- Antigen synonyms:AYTL1|1-AGP acyltransferase 11|Acetyl-CoA:lyso-PAF acetyltransferase|LysoPAFAT|AGPAT11|LPCAT-2|LPCAT2|Lysophosphatidylcholine acyltransferase 2|1-AGPAT 11|Lysophosphatidic acid acyltransferase alpha|Acyltransferase-like 1|LPC acyltransferase 2|1-acylglycerophosphocholine O-acyltransferase|1-acylglycerol-3-phosphate O-acyltransferase 11|LysoPC acyltransferase 2|1-alkylglycerophosphocholine O-acetyltransferase|Lyso-PAF acetyltransferase|Acetyl-CoA:lyso-platelet-activating factor acetyltransferase|LPAAT-alpha
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human LPCAT2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Possesses both acyltransferase and acetyltransferase activities. Activity is calcium-dependent. Involved in platelet-activating factor (PAF) biosynthesis by catalyzing the conversion of the PAF precursor, 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF) into 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine (PAF). Also converts lyso-PAF to 1-O-alkyl-2-acyl-sn-glycero-3-phosphocholine (PC), a major component of cell membranes and a PAF precursor. Under resting conditions, acyltransferase activity is preferred. Upon acute inflammatory stimulus, acetyltransferase activity is enhanced and PAF synthesis increases. Also catalyses the conversion of 1-acyl-sn-glycero-3-phosphocholine to 1,2-diacyl-sn-glycero-3-phosphocholine.
Type: Primary
Antigen: LPCAT2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/544
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat