Anti-HLA E Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1279R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HLA E
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:251-358/358
- Form:Liquid
- Gene ID:3133
- Antigen synonyms:EA2.1|HLA-E|HLA-6.2|MHC class I antigen E|EA1.2|MHC|HLA class I histocompatibility antigen, alpha chain E|HLAE|QA1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HLA E
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HLA E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA E binds a restricted subset of peptides derived from the leader peptides of other class I molecules.
Type: Primary
Antigen: HLA E
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-358/358
Host: Rabbit
Isotype: IgG
Reactivity: Human