Order Entry
Norway
ContactUsLinkComponent
 
Anti-AMPK beta 1 + 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-1116R-A750
 :  Bioss
Anti-AMPK beta 1 + 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-1116R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    AMPK beta 1 + 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Epitope:
    201-270/270
  • Form:
    Liquid
  • Gene ID:
    5564
  • Antigen synonyms:
    5'-AMP-activated protein kinase subunit beta-1|AMPK subunit beta-1|AMPK|HAMPKb|PRKAB1|AMPKb
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human AMPK beta 1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Non-catalytic subunit of AMP-activated protein kinase (AMPK), an energy sensor protein kinase that plays a key role in regulating cellular energy metabolism. In response to reduction of intracellular ATP levels, AMPK activates energy-producing pathways and inhibits energy-consuming processes: inhibits protein, carbohydrate and lipid biosynthesis, as well as cell growth and proliferation. AMPK acts via direct phosphorylation of metabolic enzymes, and by longer-term effects via phosphorylation of transcription regulators. Also acts as a regulator of cellular polarity by remodelling the actin cytoskeleton; probably by indirectly activating myosin. Beta non-catalytic subunit acts as a scaffold on which the AMPK complex assembles, via its C-terminus that bridges alpha (PRKAA1 or PRKAA2) and gamma subunits (PRKAG1, PRKAG2 or PRKAG3).

Type: Primary
Antigen: AMPK beta 1 + 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-270/270
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat