Anti-RPS6KB1 Thr 412 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5669R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RPS6KB1 Thr 412
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6198
- Antigen synonyms:p70S6K-alpha|Serine/threonine-protein kinase 14A|Ribosomal protein S6 kinase beta-1|70kDa|p70-S6K 1|PS6K|S6K-beta-1|p70 S6 Kinase|KS6B1_HUMAN|70 kDa ribosomal protein S6 kinase 1|ribosomal protein S6 kinase|P70S6K1|p70 S6K-alpha|RPS6KB1|S6K|RPS6KB1 phospho S412|p-RPS6KB1 Ser412|p70 S6 kinase alpha|p70 ribosomal S6 kinase alpha|S6K1|STK14A|Ribosomal protein S6 kinase I|p70 S6KA|P70 Beta1|p70-alpha|p70-S6K|RPS6KB1 phospho Ser412
- Modification:Phosphorylation (Thr 412 )
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:58 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human RPS6KB1 around the phosphorylation site of Thr412
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17.
Type: Primary
Antigen: RPS6KB1 Thr 412
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Others