Anti-p70 S6 kinase beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-4867R-A750
: Bioss
- Antibody type:Primary
- Antigen name:p70 S6 kinase beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:281-380/525
- Form:Liquid
- Gene ID:6199
- Antigen synonyms:P70S6K|p70S6K-alpha|Serine/threonine-protein kinase 14A|Ribosomal protein S6 kinase beta-1|p70-S6K 1|RPS6KB2|PS6K|S6K-beta-1|P70 beta 1|KS6B1_HUMAN|Serine/threonine kinase 14 alpha|Ribosomal protein S6 kinase beta 1|P70 S6 Kinase beta|70 kDa ribosomal protein S6 kinase 1|P70S6K1|p70 S6K-alpha|RPS6KB1|p70S6Kb|S6K|Ribosomal protein S6 kinase 70kDa polypeptide 2|P70 S6 Kinase|p70 alpha|p70 S6 kinase, alpha 2|p70 S6 kinase alpha|p70 ribosomal S6 kinase alpha|p70 S6K|S6K1|STK14A|P70 S6K|p70 S6 kinase, alpha 1|Ribosomal protein S6 kinase I|p70 S6KA|p70S6K alpha|p70 ribosomal S6 kinase beta 1|p70-alpha|p70-S6K|Ribosomal protein S6 kinase 70kDa polypeptide 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human p70 S6 kinase beta
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17.
Type: Primary
Antigen: p70 S6 kinase beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 281-380/525
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat