Order Entry
Norway
ContactUsLinkComponent
Anti-PKA gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-PKA gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3964R-A750
Supplier:  Bioss
Anti-PKA gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3964R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PKA gamma
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    251-351/351
  • Form:
    Liquid
  • Gene ID:
    5568
  • Antigen synonyms:
    KAPCG_HUMAN|KAPG|PRKACG|PKA C gamma|Protein kinase cAMP dependent catalytic gamma|Serine threonine protein kinase
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human PKA gamma
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

PKA (or cAPK) is a cyclic AMP dependent protein kinase. When activated by the second messenger cAMP, PKA mediates diverse cellular mechanisms, including proliferation, ion transport, regulation of metabolism, plus gene transcription. PKA is comprised of two dimers of two subunits, R (regulatory) and C (catalytic). Two families of R subunit (RI and RII) and three C subunit isoforms (C alpha, C beta, and C gamma) have been identified each possessing distinct cAMP binding properties and resulting in different phosphorylation states. C subunit is activated through autophosphorylation and direct phosphorylation at Thr197 by PDK-1. Tissue specific expression of C gamma, indicates pressure on C gamma during evolution, acting to modulate it in a functionally specific way. Certain amino acid substitutions make C gamma a distinct member of the cAMP dependent subfamily of protein kinases, and suggest that C gamma may be distinct in its protein substrate specificity or its interaction with the different regulatory subunits.

Type: Primary
Antigen: PKA gamma
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-351/351
Host: Rabbit
Isotype: IgG
Reactivity: