Anti-p70 S6 Kinase Beta Thr444 + Ser447 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3700R-A750
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:p70 S6 Kinase Beta Thr444 + Ser447
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:p70 S6 kinase beta|RPS6KB2|p70 ribosomal S6 kinase beta|S6K-beta|70 kDa ribosomal protein S6 kinase 2|P70 S6 Kinase beta phospho T444 + S447|S6K beta 2|S6K2|KS6B1_HUMAN|p70 S6KB|p70S6K beta|SRK|Ribosomal protein S6 kinase beta 2|Serine/threonine protein kinase 14 beta|p70S6Kb|S6 kinase related kinase|p70 S6Kbeta|Ribosomal protein S6 kinase 70kDa, polypeptide 2|S6K beta|p70 S6 KinaseThr444 + Ser447|p70-S6KB|Ribosomal protein S6 kinase beta-2|STK14B|p70-beta|Serine/threonine-protein kinase 14 beta|S6 kinase-related kinase|p70 beta
- Modification:Phosphorylation (Thr444 + Ser447)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human p70 S6 Kinase Beta around the phosphorylation site of Thr444/Ser447
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Spesifikasjoner
Om dette produktet
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17.
Type: Primary
Antigen: p70 S6 Kinase Beta Thr444 + Ser447
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat